Stampa…

2-Amino-4-methoxy-6-methylpyrimidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A0950-25G A0950-500G
TCIAA0950-25GEA 67 EUR
TCIAA0950-25G TCIAA0950-500G
2-Amino-4-methoxy-6-methylpyrimidine ≥98.0% (by GC, titration analysis)
2-Amino-4-methoxy-6-methylpyrimidine
Formula: C₆H₉N₃O
Peso molecolare: 139,16 g/mol
MDL Number: MFCD00059768
CAS Number: 7749-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1dba5cb77a6a70f174bf345d0f8a2ad51e0d6eb&vid=c1dba5cb77a6a70f174bf345d0f8a2ad51e0d6eb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 157.0 to 161.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR