Stampa…

3-Amino-1H-pyrazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

A1859-25G A1859-5G
TCIAA1859-25GEA 113 EUR
TCIAA1859-25G TCIAA1859-5G
3-Amino-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Amino-1H-pyrazole
Formula: C₃H₅N₃
Peso molecolare: 83,09 g/mol
Punto di ebollizione: 282°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00005236
CAS Number: 1820-80-0
EINECS: 217-338-1
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cf3fcb2ba92b001d1f194f070285cf8357a41ff&vid=3cf3fcb2ba92b001d1f194f070285cf8357a41ff&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR