Stampa…

2,6-Di-tert-butyl-p-benzoquinone ≥98.0% (by GC)

Fornitore: TCI
Attenzione

D2256-25G D2256-5G
TCIAD2256-25GEA 212 EUR
TCIAD2256-25G TCIAD2256-5G
2,6-Di-tert-butyl-p-benzoquinone ≥98.0% (by GC)
2,6-Di-tert-butyl-p-benzoquinone
Formula: C₁₄H₂₀O₂
MDL Number: MFCD00001601
CAS Number: 719-22-2
EINECS: 211-946-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ec0d8d56f8b603d884b277a62be1194337fd553&vid=4ec0d8d56f8b603d884b277a62be1194337fd553&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR