2,5-Dimethyl-p-benzoquinone ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,5-Dimethylcyclohexa-2,5-diene-1,4-dione
D0686-25G
D0686-5G
D0686-1G
TCIAD0686-25GEA
307
EUR
TCIAD0686-25G
TCIAD0686-5G
TCIAD0686-1G
2,5-Dimethyl-p-benzoquinone ≥98.0% (by GC, titration analysis)
2,5-Dimethyl-p-benzoquinone
Formula:
C₈H₈O₂ Peso molecolare: 136,15 g/mol |
MDL Number:
MFCD00041737 CAS Number: 137-18-8 EINECS: 205-283-6 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Cerium redox Method) | min. 98.0 % |
Melting point | 123.0 to 126.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...