Stampa…

3-Acetylcoumarin ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 3-Acetyl-2H-chromen-2-one

A2200-25G A2200-5G
TCIAA2200-25GEA 119 EUR
TCIAA2200-25G TCIAA2200-5G
3-Acetylcoumarin ≥98.0% (by GC)
3-Acetylcoumarin
Formula: C₁₁H₈O₃
Peso molecolare: 188,18 g/mol
MDL Number: MFCD00006853
CAS Number: 3949-36-8
EINECS: 223-541-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5521f47ce156cd84cdfbb60d39d30b3a30b2cd57&vid=5521f47ce156cd84cdfbb60d39d30b3a30b2cd57&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR