Stampa…

3-Chlorocoumarin ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C2297-1G C2297-5G
TCIAC2297-1GEA 44 EUR
TCIAC2297-1G TCIAC2297-5G
3-Chlorocoumarin ≥98.0% (by GC)
3-Chlorocoumarin
Formula: C₉H₅ClO₂
Peso molecolare: 180,59 g/mol
MDL Number: MFCD00024073
CAS Number: 92-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a74a38649e8313e5a5a86bc30df5857607983890&vid=a74a38649e8313e5a5a86bc30df5857607983890&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR