2,2,6,6-Tetramethylpiperidin-4-ol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine
T0910-25G
T0910-500G
TCIAT0910-25GEA
26
EUR
TCIAT0910-25G
TCIAT0910-500G
2,2,6,6-Tetramethylpiperidin-4-ol ≥98.0% (by GC, titration analysis)
2,2,6,6-Tetramethylpiperidin-4-ol
Formula:
C₉H₁₉NO Peso molecolare: 157,26 g/mol |
MDL Number:
MFCD00005983 CAS Number: 2403-88-5 EINECS: 219-291-2 UN: 3259 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 130.0 to 133.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...