Stampa…

4-Amino-1-benzylpiperidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Amino-N-benzylpiperidine

A1086-25G A1086-500G A1086-100G
TCIAA1086-25GEA 84 EUR
TCIAA1086-25G TCIAA1086-500G TCIAA1086-100G
4-Amino-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
4-Amino-1-benzylpiperidine
Formula: C₁₂H₁₈N₂
Peso molecolare: 190,29 g/mol
MDL Number: MFCD00006504
CAS Number: 50541-93-0
EINECS: 256-620-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce19b8dd1f13a0db0f343401599f2358ad156de&vid=8ce19b8dd1f13a0db0f343401599f2358ad156de&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR