Stampa…

(Phenylthio)trimethylsilane ≥95.0% (by GC)

Fornitore: TCI
Attenzione

P1378-25G P1378-5G
TCIAP1378-25GEA 272 EUR
TCIAP1378-25G TCIAP1378-5G
(Phenylthio)trimethylsilane ≥95.0% (by GC)
(Phenylthio)trimethylsilane
Formula: C₉H₁₄SSi
Punto di ebollizione: 65°C
Flash Pt: 33°C
MDL Number: MFCD00008272
CAS Number: 4551-15-9
EINECS: 224-916-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4be418c0d2e0b6063f75bcfe5e8f5fe066876ec&vid=c4be418c0d2e0b6063f75bcfe5e8f5fe066876ec&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR