Stampa…

Thiazolidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

T0793-25G T0793-5G
TCIAT0793-25GEA 165 EUR
TCIAT0793-25G TCIAT0793-5G
Thiazolidine ≥98.0% (by GC, titration analysis)
Thiazolidine
Formula: C₃H₇NS
Peso molecolare: 89,16 g/mol
Punto di ebollizione: 165°C
Flash Pt: 56°C
MDL Number: MFCD00005211
CAS Number: 504-78-9
EINECS: 208-002-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=728983b2e73533621234d93dd27e8497a00a7e0d&vid=728983b2e73533621234d93dd27e8497a00a7e0d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR