Stampa…

Ethyl isobutyrylacetate ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 4-methyl-3-oxopentanoate

E0882-25G E0882-5G
TCIAE0882-25GEA 118 EUR
TCIAE0882-25G TCIAE0882-5G
Ethyl isobutyrylacetate ≥95.0% (by GC)
Ethyl isobutyrylacetate
Formula: C₈H₁₄O₃
Peso molecolare: 158,2 g/mol
Punto di ebollizione: 90°C
Flash Pt: 44°C
MDL Number: MFCD00009198
CAS Number: 7152-15-0
EINECS: 230-491-9
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21566831159f30757304db6c2f359617e2f98b1&vid=b21566831159f30757304db6c2f359617e2f98b1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR