Sodium-tert-butanolate ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate
S0450-100G
S0450-25G
S0450-500G
TCIAS0450-100GEA
31
EUR
TCIAS0450-100G
TCIAS0450-25G
TCIAS0450-500G
Sodium-tert-butanolate ≥98.0% (by titrimetric analysis)
Sodium-tert-butanolate
Formula:
C₄H₉NaO Peso molecolare: 96,1 g/mol |
MDL Number:
MFCD00040575 CAS Number: 865-48-5 EINECS: 212-741-9 UN: 3206 ADR: 4.2,II |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...