2,5-Dichloro-p-benzoquinone ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₂Cl₂O₂ |
MDL Number:
MFCD00041736 CAS Number: 615-93-0 EINECS: 210-453-8 |
|
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...