Stampa…

Methyl butyrylacetate ≥96.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 3-oxohexanoate

K0037-25ML K0037-500ML
TCIAK0037-25MLEA 18 EUR
TCIAK0037-25ML TCIAK0037-500ML
Methyl butyrylacetate ≥96.0% (by GC)
Methyl butyrylacetate
Formula: C₇H₁₂O₃
Peso molecolare: 144,17 g/mol
Punto di ebollizione: 85°C
Flash Pt: 90°C
MDL Number: MFCD00040995
CAS Number: 30414-54-1
EINECS: 250-185-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=610b988711b84c145f1efbd35ac3dd6d4b1ff9fa&vid=610b988711b84c145f1efbd35ac3dd6d4b1ff9fa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR