Ethyl butyrylacetate ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethyl Butyrylacetate, Ethyl 3-oxohexanoate
K0030-25ML
K0030-500ML
TCIAK0030-25MLEA
22
EUR
TCIAK0030-25ML
TCIAK0030-500ML
Ethyl butyrylacetate ≥95.0% (by GC)
Ethyl butyrylacetate
Formula:
C₈H₁₄O₃ Peso molecolare: 158,2 g/mol Punto di ebollizione: 95°C Flash Pt: 78°C |
MDL Number:
MFCD00009401 CAS Number: 3249-68-1 EINECS: 221-835-9 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...