3-Methyl-1H-pyrazole ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
3-Methylpyrazole
M0564-25G
M0564-5G
TCIAM0564-25GEA
67
EUR
TCIAM0564-25G
TCIAM0564-5G
3-Methyl-1H-pyrazole ≥97.0% (by GC)
3-Methyl-1H-pyrazole
Formula:
C₄H₆N₂ Punto di ebollizione: 204°C Flash Pt: 93°C |
MDL Number:
MFCD00005240 CAS Number: 1453-58-3 EINECS: 215-925-7 UN: 2810 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...