Stampa…

Elaidic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (E)-Octadec-9-enoic acid

O0010-25G O0010-5G
TCIAO0010-25GEA 321 EUR
TCIAO0010-25G TCIAO0010-5G
Elaidic acid ≥97.0% (by titrimetric analysis)
Elaidic acid
Formula: C₁₈H₃₄O₂
Peso molecolare: 282,47 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00063954
CAS Number: 112-79-8
EINECS: 204-006-6
Merck Index: 14,03533
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee1d8f74c4ab92ffa45d73a5137fe6828bd66948&vid=ee1d8f74c4ab92ffa45d73a5137fe6828bd66948&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR