Stampa…

Tetrachlorohydroquinone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1,2,4,5-Tetrachloro-3,6-dihydroxybenzene, Tetrachloroquinol

T0067-25G T0067-5G
TCIAT0067-25GEA 93 EUR
TCIAT0067-25G TCIAT0067-5G
Tetrachlorohydroquinone ≥98.0% (by GC)
Tetrachlorohydroquinone
Formula: C₆H₂Cl₄O₂
Peso molecolare: 247,89 g/mol
MDL Number: MFCD00041748
CAS Number: 87-87-6
EINECS: 201-779-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=332efe82ac95eb3968b12001dad35a178c0ef7e4&vid=332efe82ac95eb3968b12001dad35a178c0ef7e4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR