Stampa…

Bromohydroquinone ≥90.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromobenzene-1,4-diol

B1600-25G B1600-5G
TCIAB1600-25GEA 545 EUR
TCIAB1600-25G TCIAB1600-5G
Bromohydroquinone ≥90.0% (by GC)
Bromohydroquinone
Formula: C₆H₅BrO₂
Peso molecolare: 189,01 g/mol
MDL Number: MFCD00041747
CAS Number: 583-69-7
EINECS: 209-516-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f47a1c33871eb17687c7d47e3846b5576c88f29&vid=1f47a1c33871eb17687c7d47e3846b5576c88f29&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 90.0 %
Melting point 112.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR