Stampa…

9-Methylacridine ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

M1237-1G M1237-5G
TCIAM1237-1GEA 93 EUR
TCIAM1237-1G TCIAM1237-5G
9-Methylacridine ≥97.0% (by GC)
9-Methylacridine
Formula: C₁₄H₁₁N
Peso molecolare: 193,25 g/mol
MDL Number: MFCD00143523
CAS Number: 611-64-3
EINECS: 210-272-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec5a2b0a358dfd4ad3d5df6367ba5e7f28f8278&vid=8ec5a2b0a358dfd4ad3d5df6367ba5e7f28f8278&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 97.0 %
Melting point 116.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR