Stampa…

1-Butyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: N-Butyl-N-methylpyrrolidinium bromide

B3427-25G B3427-5G
TCIAB3427-25GEA 108 EUR
TCIAB3427-25G TCIAB3427-5G
1-Butyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)
1-Butyl-1-methylpyrrolidinium bromide
Formula: C₉H₂₀BrN
Peso molecolare: 222,17 g/mol
MDL Number: MFCD07784439
CAS Number: 93457-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efd73e1f0cf3e4b33b7c990a6cf91237b3f71086&vid=efd73e1f0cf3e4b33b7c990a6cf91237b3f71086&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR