Stampa…

o-Phenylenediacetonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1,2-Phenylenediacetonitrile, 1,2-Bis(cyanomethyl)-benzene

X0050-25G X0050-5G
TCIAX0050-25GEA 77 EUR
TCIAX0050-25G TCIAX0050-5G
o-Phenylenediacetonitrile ≥98.0% (by GC)
o-Phenylenediacetonitrile
Formula: C₁₀H₈N₂
Peso molecolare: 156,19 g/mol
MDL Number: MFCD00001905
CAS Number: 613-73-0
EINECS: 210-351-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d09bbe2536b5eb3062cf3e6554feb4273a5cdd89&vid=d09bbe2536b5eb3062cf3e6554feb4273a5cdd89&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR