Stampa…

4-Aminoazobenzene ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: 4-Aminoazobenzene, p-Aminoazobenzene, 4-(Phenylazo)aniline, 4-Phenylazoaniline

P0141-25G P0141-100G
TCIAP0141-25GEA 35 EUR
TCIAP0141-25G TCIAP0141-100G
4-Aminoazobenzene ≥98.0% (by HPLC, titration analysis)
4-Aminoazobenzene
Formula: C₁₂H₁₁N₃
MDL Number: MFCD00007854
CAS Number: 60-09-3
EINECS: 200-453-6
UN: 3077
ADR: 9,III
Merck Index: 14,00419
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c2ebad3d08020f3177190e70be933c1ce0797a1&vid=4c2ebad3d08020f3177190e70be933c1ce0797a1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR