S-Propyl thioacetate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
S-Propyl ethanethioate
P1716-25G
P1716-5G
TCIAP1716-25GEA
55
EUR
TCIAP1716-25G
TCIAP1716-5G
S-Propyl thioacetate ≥98.0% (by GC)
S-Propyl thioacetate
Formula:
C₅H₁₀OS Peso molecolare: 118,2 g/mol Punto di ebollizione: 137°C |
MDL Number:
MFCD00039937 CAS Number: 2307-10-0 EINECS: 218-984-7 UN: 3272 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...