Stampa…

Ethyl carbazate ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Ethyl hydrazinecarboxylate

C1097-25G C1097-500G
TCIAC1097-25GEA 58 EUR
TCIAC1097-25G TCIAC1097-500G
Ethyl carbazate ≥99.0% (by titrimetric analysis)
Ethyl carbazate
Formula: C₃H₈N₂O₂
Peso molecolare: 104,11 g/mol
Punto di ebollizione: 118°C
MDL Number: MFCD00007595
CAS Number: 4114-31-2
EINECS: 223-903-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5774f9936bc430922668d36e4283c27f1baf946&vid=f5774f9936bc430922668d36e4283c27f1baf946&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 44.0 to 48.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR