Stampa…

Methyl oxalyl chloride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate

C1561-25G C1561-250G
TCIAC1561-25GEA 76 EUR
TCIAC1561-25G TCIAC1561-250G
Methyl oxalyl chloride ≥98.0% (by GC, titration analysis)
Methyl oxalyl chloride
Formula: C₃H₃ClO₃
Peso molecolare: 122,51 g/mol
Punto di ebollizione: 125°C
Flash Pt: 46°C
MDL Number: MFCD00000705
CAS Number: 5781-53-3
EINECS: 227-307-4
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a4ad9f806f8ea16030c093adc3b8ae2bf9d06fa&vid=6a4ad9f806f8ea16030c093adc3b8ae2bf9d06fa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR