(±)-2-Bromopropionyl bromide ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
B0649-25G
B0649-500G
B0649-100G
TCIAB0649-25GEA
16
EUR
TCIAB0649-25G
TCIAB0649-500G
TCIAB0649-100G
(±)-2-Bromopropionyl bromide ≥98.0% (by titrimetric analysis)
(±)-2-Bromopropionyl bromide
Formula:
C₃H₄Br₂O Punto di ebollizione: 155°C Flash Pt: 109°C |
MDL Number:
MFCD00000142 CAS Number: 563-76-8 EINECS: 209-261-7 UN: 3265 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...