Stampa…

Dimethoxy(methyl)silane ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

D2100-25ML D2100-100ML
TCIAD2100-25MLEA 94 EUR
TCIAD2100-25ML TCIAD2100-100ML
Dimethoxy(methyl)silane ≥97.0% (by GC)
Dimethoxy(methyl)silane
Formula: C₃H₁₀O₂Si
Punto di ebollizione: 61°C
Flash Pt: -9°C
MDL Number: MFCD00048019
CAS Number: 16881-77-9
EINECS: 240-914-9
UN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79aaa4aa107c4a3eb59ce698c83a491e89434e40&vid=79aaa4aa107c4a3eb59ce698c83a491e89434e40&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR