(±)-Methyl-2-bromopropionate ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₄H₇BrO₂ Punto di ebollizione: 144°C Flash Pt: 51°C |
MDL Number:
MFCD00000143 CAS Number: 5445-17-0 EINECS: 226-642-3 UN: 2920 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...