(±)-Ethyl-2-bromopropionate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate
B0646-25G
B0646-500G
B0646-100G
TCIAB0646-25GEA
24
EUR
TCIAB0646-25G
TCIAB0646-500G
TCIAB0646-100G
(±)-Ethyl-2-bromopropionate ≥98.0% (by GC)
(±)-Ethyl-2-bromopropionate
Formula:
C₅H₉BrO₂ Peso molecolare: 181,03 g/mol Punto di ebollizione: 160°C Flash Pt: 51°C |
MDL Number:
MFCD00000144 CAS Number: 535-11-5 EINECS: 208-609-5 UN: 1992 ADR: 3,III Merck Index: 14,03772 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...