Stampa…

1,4-Diformylpiperazine

Fornitore: TCI

D2578-25G D2578-500G D2578-100G
TCIAD2578-25GEA 47 EUR
TCIAD2578-25G TCIAD2578-500G TCIAD2578-100G
1,4-Diformylpiperazine
1,4-Diformylpiperazine
Formula: C₆H₁₀N₂O₂
Peso molecolare: 142.16 g/mol
MDL Number: MFCD00006153
CAS Number: 4164-39-0
EINECS: 224-011-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5de19ee70f2205a3bcd063dd3b3e1cb60d48d9eb&vid=5de19ee70f2205a3bcd063dd3b3e1cb60d48d9eb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 129.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR