Stampa…

Isopentyl propionate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Isoamyl Propionate, Propionic acid isoamyl ester

I0677-25G I0677-500G
TCIAI0677-25GEA 16 EUR
TCIAI0677-25G TCIAI0677-500G
Isopentyl propionate ≥98.0% (by GC)
Isopentyl propionate
Formula: C₈H₁₆O₂
Peso molecolare: 144,21 g/mol
Punto di ebollizione: 161°C
Flash Pt: 41°C
MDL Number: MFCD00048611
CAS Number: 105-68-0
EINECS: 203-322-1
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b10d3de321f1a08d2ca633aaedaeebaf97f013b5&vid=b10d3de321f1a08d2ca633aaedaeebaf97f013b5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR