Stampa…

Methyl propionylacetate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate

K0035-25G K0035-500G K0035-100G
TCIAK0035-25GEA 24 EUR
TCIAK0035-25G TCIAK0035-500G TCIAK0035-100G
Methyl propionylacetate ≥98.0% (by GC)
Methyl propionylacetate
Formula: C₆H₁₀O₃
Peso molecolare: 130,14 g/mol
Punto di ebollizione: 74°C
Flash Pt: 71°C
MDL Number: MFCD00011705
CAS Number: 30414-53-0
EINECS: 250-184-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb9776634625841668d8cd7185bba962be632f49&vid=eb9776634625841668d8cd7185bba962be632f49&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR