Stampa…

Tris(2-ethylhexyl)phosphate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Trioctyl phosphate, Phosphoric acid tris(2-ethylhexyl ester), Tris(2-ethylhexyl) phosphate

P1022-25ML P1022-500ML
TCIAP1022-25MLEA 16 EUR
TCIAP1022-25ML TCIAP1022-500ML
Tris(2-ethylhexyl)phosphate ≥98.0% (by GC)
Tris(2-ethylhexyl)phosphate
Formula: C₂₄H₅₁O₄P
Peso molecolare: 434,64 g/mol
Punto di ebollizione: 215°C
Flash Pt: 204°C
MDL Number: MFCD00009491
CAS Number: 78-42-2
EINECS: 201-116-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=267c04cdc182bcfaf0b72b124da3eb883e926b2a&vid=267c04cdc182bcfaf0b72b124da3eb883e926b2a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR