1-Chloroadamantane ≥98.0% (by GC)
Fornitore: TCI EUROPE
C0747-25G
C0747-5G
TCIAC0747-25GEA
105
EUR
TCIAC0747-25G
TCIAC0747-5G
1-Chloroadamantane ≥98.0% (by GC)
1-Chloroadamantane
Formula:
C₁₀H₁₅Cl Peso molecolare: 170,68 g/mol Punto di fusione: 163…166 °C Densità: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00075627 CAS Number: 935-56-8 EINECS: 213-305-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...