Stampa…

2-Chloro-6-ethoxypyridine ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C1143-25G C1143-5G
TCIAC1143-25GEA 136 EUR
TCIAC1143-25G TCIAC1143-5G
2-Chloro-6-ethoxypyridine ≥97.0% (by GC)
2-Chloro-6-ethoxypyridine
Formula: C₇H₈ClNO
Peso molecolare: 157,6 g/mol
MDL Number: MFCD00023465
CAS Number: 42144-78-5
EINECS: 255-681-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1e1693caecc3ef46b94ee3ef6792d55784a85d9&vid=b1e1693caecc3ef46b94ee3ef6792d55784a85d9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR