Stampa…

1,4-Naphthalenedicarboxylic acid ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Naphthalene-1,4-dicarboxylic acid

N0606-25G N0606-100G
TCIAN0606-25GEA 55 EUR
TCIAN0606-25G TCIAN0606-100G
1,4-Naphthalenedicarboxylic acid ≥95.0% (by titrimetric analysis)
1,4-Naphthalenedicarboxylic acid
Formula: C₁₂H₈O₄
Peso molecolare: 216,19 g/mol
MDL Number: MFCD00014312
CAS Number: 605-70-9
EINECS: 210-094-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ddb454e5f30b38b71a584d2a22b0b3bc433f2a0&vid=6ddb454e5f30b38b71a584d2a22b0b3bc433f2a0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR