Stampa…

5-Bromoisoquinoline ≥96.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B3313-1G B3313-5G
TCIAB3313-1GEA 16 EUR
TCIAB3313-1G TCIAB3313-5G
5-Bromoisoquinoline ≥96.0% (by GC, titration analysis)
5-Bromoisoquinoline
Formula: C₉H₆BrN
Punto di ebollizione: 97°C
MDL Number: MFCD01646405
CAS Number: 34784-04-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27572b252c4366e2c4a4e8ccacc2df1b166cb256&vid=27572b252c4366e2c4a4e8ccacc2df1b166cb256&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR