Methyl-2-aminonicotinate ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2-aminonicotinate
M2125-1G
M2125-5G
TCIAM2125-1GEA
21
EUR
TCIAM2125-1G
TCIAM2125-5G
Methyl-2-aminonicotinate ≥97.0% (by GC, titration analysis)
Methyl-2-aminonicotinate
Formula:
C₇H₈N₂O₂ Peso molecolare: 152,15 g/mol |
MDL Number:
MFCD00128872 CAS Number: 14667-47-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...