5-Chloro-2-hydroxy-3-nitropyridine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Chloro-3-nitropyridin-2-ol
C1972-25G
C1972-5G
TCIAC1972-25GEA
266
EUR
TCIAC1972-25G
TCIAC1972-5G
5-Chloro-2-hydroxy-3-nitropyridine ≥98.0% (by titrimetric analysis)
5-Chloro-2-hydroxy-3-nitropyridine
Formula:
C₅H₃ClN₂O₃ Peso molecolare: 174,54 g/mol |
MDL Number:
MFCD00114884 CAS Number: 21427-61-2 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 93.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...