2,6-Dichloro-5-fluoronicotinic acid ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,6-Dichloro-5-fluoropyridine-3-carboxylic acid
D2610-25G
D2610-5G
TCIAD2610-25GEA
45
EUR
TCIAD2610-25G
TCIAD2610-5G
2,6-Dichloro-5-fluoronicotinic acid ≥97.0% (by titrimetric analysis)
2,6-Dichloro-5-fluoronicotinic acid
Formula:
C₆H₂Cl₂FNO₂ Peso molecolare: 209,99 g/mol |
MDL Number:
MFCD00799517 CAS Number: 82671-06-5 |
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 151.0 to 156.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...