Stampa…

Methyl-6-chloronicotinate ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 6-chloronicotinate

M1980-25G M1980-5G
TCIAM1980-25GEA 114 EUR
TCIAM1980-25G TCIAM1980-5G
Methyl-6-chloronicotinate ≥98.0% (by GC, total nitrogen)
Methyl-6-chloronicotinate
Formula: C₇H₆ClNO₂
Peso molecolare: 171,58 g/mol
MDL Number: MFCD00023420
CAS Number: 73781-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10f06c7f5aa61185548b58606ec8671757e1524b&vid=10f06c7f5aa61185548b58606ec8671757e1524b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR