5-Bromo-2-hydroxy-3-nitropyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Bromo-3-nitro-2-pyridinol, 5-Bromo-3-nitropyridin-2(1H)-one
B2706-1G
B2706-5G
TCIAB2706-1GEA
16
EUR
TCIAB2706-1G
TCIAB2706-5G
5-Bromo-2-hydroxy-3-nitropyridine ≥98.0% (by GC, titration analysis)
5-Bromo-2-hydroxy-3-nitropyridine
Formula:
C₅H₃BrN₂O₃ Peso molecolare: 218,99 g/mol Punto di fusione: 247°C |
MDL Number:
MFCD00023473 CAS Number: 15862-34-7 EINECS: 239-989-0 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...