Phenyl dichlorophosphate ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₆H₅Cl₂O₂P Punto di ebollizione: 240°C |
MDL Number:
MFCD00002067 CAS Number: 770-12-7 EINECS: 212-220-6 UN: 3265 ADR: 8,II Merck Index: 14,07282 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...