3-Methyl-1-phenyl-2-phospholene-1-oxide ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
4-Methyl-1-phenyl-2,3-dihydro-1H-phosphole 1-oxide
M0705-25G
M0705-5G
TCIAM0705-25GEA
208
EUR
TCIAM0705-25G
TCIAM0705-5G
3-Methyl-1-phenyl-2-phospholene-1-oxide ≥95.0% (by HPLC)
3-Methyl-1-phenyl-2-phospholene-1-oxide
Formula:
C₁₁H₁₃OP Peso molecolare: 192,2 g/mol Punto di ebollizione: 155°C |
MDL Number:
MFCD00014518 CAS Number: 707-61-9 EINECS: 211-901-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...