Stampa…

Diphenyliodonium nitrate

Fornitore: TCI
Attenzione

D2357-25G D2357-5G
TCIAD2357-25GEA 192 EUR
TCIAD2357-25G TCIAD2357-5G
Diphenyliodonium nitrate
Diphenyliodonium nitrate
Formula: C₁₂H₁₀INO₃
MDL Number: MFCD00031704
CAS Number: 722-56-5
EINECS: 211-962-8
UN: 1479
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f8be15b0d8799d51e9d1aa79bb161e6df8238fc&vid=0f8be15b0d8799d51e9d1aa79bb161e6df8238fc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR