1-Phenyl-2-(trimethylsilyl)acetylene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Trimethyl(phenylethynyl)silane
P1398-5ML
P1398-25ML
TCIAP1398-5MLEA
32
EUR
TCIAP1398-5ML
TCIAP1398-25ML
1-Phenyl-2-(trimethylsilyl)acetylene ≥98.0% (by GC)
1-Phenyl-2-(trimethylsilyl)acetylene
Formula:
C₁₁H₁₄Si Peso molecolare: 174,32 g/mol Flash Pt: 73°C |
MDL Number:
MFCD00054860 CAS Number: 2170-06-1 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...