Stampa…

4H-3,1-Benzossazin-2,4(1H)-dione ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1H-Benzo[d][1,3]oxazine-2,4-dione

I0081-25G I0081-500G
TCIAI0081-25GEA 49 EUR
TCIAI0081-25G TCIAI0081-500G
4H-3,1-Benzossazin-2,4(1H)-dione ≥98.0% (by HPLC, titration analysis)
4H-3,1-Benzossazin-2,4(1H)-dione
Formula: C₈H₅NO₃
Peso molecolare: 163,13 g/mol
MDL Number: MFCD00006700
CAS Number: 118-48-9
EINECS: 204-255-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d5c717290feb66f585af5e77501e8a7a22890ef&vid=7d5c717290feb66f585af5e77501e8a7a22890ef&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR