2-Mercapto-2-thiazoline ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol
M0065-25G
M0065-500G
TCIAM0065-25GEA
16
EUR
TCIAM0065-25G
TCIAM0065-500G
2-Mercapto-2-thiazoline ≥98.0% (by titrimetric analysis)
2-Mercapto-2-thiazoline
Formula:
C₃H₅NS₂ Peso molecolare: 119,21 g/mol |
MDL Number:
MFCD00126013 CAS Number: 96-53-7 EINECS: 202-512-1 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 104.0 to 107.0 deg-C |
Solubility in hot Water | almost transparency |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...