Iodosobenzene diacetate ≥97.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Iodobenzene diacetate, (Diacetoxyiodo)benzene
Formula:
C₁₀H₁₁IO₄ Peso molecolare: 322.1 g/mol |
MDL Number:
MFCD00008692 CAS Number: 3240-34-4 EINECS: 221-808-1 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...